missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26208-1-AP-20UL
This item is not returnable.
View return policy
Description
CENPK is a subunit of a CENPH -CENPI -associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.Specifications
| CENPK | |
| Polyclonal | |
| Unconjugated | |
| CENPK | |
| AF5alpha, CENP K, CENPK, centromere protein K, FKSG14, ICEN37, P33, Protein AF 5alpha, Solt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64105 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BS16 | |
| CENPK | |
| CENPK Fusion Protein Ag23823 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction