Learn More
CENPO Monoclonal Antibody (OTI7F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809481S
This item is not returnable.
View return policy
Description
CENPO Monoclonal antibody specifically detects CENPO in Human samples. It is validated for Western BlotSpecifications
| CENPO | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P11766 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 128 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI7F9 | |
| Unconjugated | |
| ADH5 | |
| 2810429O05Rik; 8430427C03Rik; CENPO; CENP-O; Centromere protein O; centromeric protein O; D12Ertd482e; ICEN36; ICEN-36; Interphase centromere complex protein 36; MCM21R | |
| Human recombinant protein fragment corresponding to amino acids 1-266 of human ADH5 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |