missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPP Antibody (3G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00401541-M07
This item is not returnable.
View return policy
Description
CENPP Monoclonal antibody specifically detects CENPP in Human samples. It is validated for Western Blot, ELISA
Specifications
| CENPP | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| CENP-PRP11-19J3.3, centromere protein P, FLJ33928 | |
| CENPP (NP_001012267, 1 a.a. ∽ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDAELAEVRALQAEIAALRRACEDPPAPWEEKSRVQKSFQAIHQFNLEGWKSSKDLKNQLGHLESELSFLSTLTGINIRNHSKQTEDLTSTEMTEKSI | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 401541 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 3G8 | |
| Western Blot 1:500, ELISA | |
| NP_001012267 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction