missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPQ Antibody (OTI1C5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71991CL1
This item is not returnable.
View return policy
Description
CENPQ Monoclonal antibody specifically detects CENPQ in Human samples. It is validated for Western Blot
Specifications
| CENPQ | |
| Monoclonal | |
| CoraFluor 1 | |
| C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139 | |
| Full length human recombinant protein of human CENPQ (NP_060602) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 55166 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| OTI1C5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction