missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPQ Antibody (OTI1C5), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71991C
This item is not returnable.
View return policy
Description
CENPQ Monoclonal specifically detects CENPQ in Human samples. It is validated for Western Blot.
Specifications
| CENPQ | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CENPQ | |
| Full length recombinant protein of human CENPQ (NP_060602) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI1C5 | |
| Western Blot | |
| C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 55166.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction