missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPQ Antibody (OTI1C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46240
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
CENPQ Monoclonal specifically detects CENPQ in Human samples. It is validated for Western Blot.
Specifications
| CENPQ | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| C6orf139, CENP-QFLJ10545, centromere protein Q, chromosome 6 open reading frame 139 | |
| Mouse | |
| 30.4 kDa | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 55166 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CENPQ | |
| Full length human recombinant protein of human CENPQ (NP_060602) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction