missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENTD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92070-0.1ml
This item is not returnable.
View return policy
Description
CENTD1 Polyclonal antibody specifically detects CENTD1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CENTD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Arf and Rho GAP adapter protein 2, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2, centaurin, delta 1, Centaurin-delta-1, CENTD1arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2, cnt-d1, FLJ13675, FLJ44916, KIAA0580centaurin-delta-1, PARX, Protein PARX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1405-1704 of human ARAP2 (NP_056045.2). LAEPGSAYLVVKRFLTADTIKHCSDRSTLGSIKEGILKIKEEPSKILSGNKFQDRYFVLRDGFLFLYKDVKSSKHDKMFSLSSMKFYRGVKKKMKPPTSWGLTAYSEKHHWHLCCDSSRTQTEWMTSIFIAQHEYDIWPPAGKERKRSITKNPKIGGLPLIPIQHEGNATLARKNIESARAELERLRLSEKCDKESVDSSLKERASMVAHCLEHKDDKLRNRPRKHRSFNCLEDTEPEAPLGQPKGHKGLKTLRKTEDRNSKATLDSDHKLPSRVIEELNVVLQRSRTLPKELQDEQILK | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 116984 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction