Learn More
Grant Instruments Centrifuge Buckets
Product Code. 30274341
Change view
Click to view available options
Tube Size:
2 to 5 mL Vacutainers
4 to 8 mL Vacutainers
8 to 10 mL Vacutainers
Unit Size:
Pack of 24
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Size unitSize
30274341 4 to 8 mL Vacutainers Pack of 24
30274342 2 to 5 mL Vacutainers Pack of 24
30274350 8 to 10 mL Vacutainers Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30274341 Supplier Grant Instruments Supplier No. BN13/10024
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
For rotors R-24/10, Ri-24/10 optional adapter
  • Buckets for Ri-24/10 rotor
  • Set of 24 pcs.
  • For use with LMC-56
Available sizes:
  • 13 x 100 mm tubes (e.g.4 to 8 mL vacutainers)
  • 13 x 75 mm tubes (e.g.2 to 5 mL vacutainers)
  • 16 x 100 mm tubes (e.g.8 to 9 mL vacutainers)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LMC-56, R-24/10, Ri-24/10
Type Bucket
Bore Size (Metric) 16 mm
Color White
Container Type Vacutainer Adaptor
Material Engineered Polymer
No. of Places 1
Quantity 24/Pk.
Tube Size 4 to 8 mL Vacutainers
Tube Type Vacutainers