missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Centrin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104315
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Centrin 1.
The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
Specifications
| Centrin 1 | |
| Polyclonal | |
| Unconjugated | |
| CETN1 | |
| calcium binding protein; Calt; Caltractin; Caltractin isoform 2; CEN1; centrin 1; centrin 1 S homeolog; centrin, EF-hand protein, 1; centrin, EF-hand protein, 2; centrin-1; CETN; cetn1; cetn1.S; cetn2; EF-hand protein; testicular tissue protein Li 37; testis-specific murine centrin; Unknown (protein for MGC:133570); xcen; XELAEV_18042289mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1068, 26369, 84592 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41209, Q12798 | |
| CETN1 | |
| A synthesized peptide derived from human CETN1(Accession Q12798), corresponding to amino acid residues T94-E144. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction