missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Centrin 3 Antibody (OTI4G5), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70409AF750
This item is not returnable.
View return policy
Description
Centrin 3 Monoclonal specifically detects Centrin 3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Centrin 3 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CETN3 | |
| Full length recombinant protein of human Centrin 3 (NP_004356) produced in E.coli. | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot | |
| OTI4G5 | |
| Western Blot | |
| CDC31 yeast homolog, CEN3centrin, EF-hand protein, 3 (CDC31 homolog, yeast), centrin, EF-hand protein, 3, centrin, EF-hand protein, 3 (CDC31 yeast homolog), centrin-3, EF-hand superfamily member, MGC12502, MGC138245 | |
| Mouse | |
| 0.1 mL | |
| 1070 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction