missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP120 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570741
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
This gene encodes a protein that functions in the microtubule-dependent coupling of the nucleus and the centrosome. A similar protein in mouse plays a role in both interkinetic nuclear migration, which is a characteristic pattern of nuclear movement in neural progenitors, and in neural progenitor self-renewal. Mutations in this gene are predicted to result in neurogenic defects. Alternative splicing results in multiple transcript variants.
Specifications
| CEP120 | |
| Polyclonal | |
| Unconjugated | |
| CEP120 | |
| A230075C01; AU016693; Ccdc100; centrosomal protein 120; centrosomal protein 120kDa; centrosomal protein of 120 kDa; Cep120; coiled-coil domain containing 100; coiled-coil domain-containing protein 100; RGD1565619; SRTD13 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 153241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N960 | |
| CEP120 | |
| synthetic peptide directed towards the N-terminal region of human CEP120. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction