missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP128 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78745UV
This item is not returnable.
View return policy
Description
CEP128 Polyclonal antibody specifically detects CEP128 in Human samples. It is validated for Western Blot
Specifications
| CEP128 | |
| Polyclonal | |
| Western Blot | |
| centrosomal protein 128kDa | |
| The immunogen for this product maps to a region between residue 1044 and 1094 of human Centrosomal Protein 128kDa using the numbering given in entry NP_689659.2 (GeneID 145508). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 145508 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction