Learn More
CEP164 Antibody (13), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-43631
Product Code. 18748803
  • € 590.00 / 0.10mL
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
CEP164 Monoclonal specifically detects CEP164 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse
The immunogen used to generate this antibody corresponds to human CEP164.
0.1 mL