missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP170 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55317
This item is not returnable.
View return policy
Description
CEP170 Polyclonal specifically detects CEP170 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation Sequencing.
Specifications
| CEP170 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| centrosomal protein 170kDa, Cep170, KARP 1 binding protein, KARP-1-binding protein, KARP1-binding protein, KIAA0470FAM68AKABcentrosomal protein of 170 kDa, XRCC5 binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9859 | |
| Human | |
| IgG |
| ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunofluorescence, Chromatin Immunoprecipitation Sequencing | |
| Q5SW79 | |
| CEP170 | |
| The immunogen recognized by this antibody maps to a region between residue 1534 and the C-teriminus (residue 1584) of human centrosomal protein 170kDa using the numbering given in entry NP_055627.2 (GeneID 9859). | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Chimpanzee. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction