missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP192 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28718AF700
This item is not returnable.
View return policy
Description
CEP192 Polyclonal antibody specifically detects CEP192 in Human samples. It is validated for Western Blot
Specifications
| CEP192 | |
| Polyclonal | |
| Western Blot | |
| centrosomal protein 192kDa, centrosomal protein of 192 kDa, Cep192, FLJ10352, KIAA1569 | |
| The immunogen recognized by this antibody maps to a region between residue 1800 and 1850 of human centrosomal protein using the numbering given in entry NP_115518.2 (GeneID 55125). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55125 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction