missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP192 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CEP192 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17211416
|
Proteintech
28700-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17201416
|
Proteintech
28700-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CEP192 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CEP192. This antibody recognizes human antigen. The CEP192 antibody has been validated for the following applications: IF, WB,ELISA analysis.
Specifications
| CEP192 | |
| Western Blot, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| 55125 |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title