missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP250/CNAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14498-1-AP-150UL
This item is not returnable.
View return policy
Description
Probably plays an important role in centrosome cohesion during interphase.Specifications
| CEP250/CNAP1 | |
| Polyclonal | |
| Unconjugated | |
| CEP250 | |
| 250 kDa centrosomal protein, C NAP1, Centrosomal protein 2, centrosomal protein 250kDa, CEP2, CEP250, CNAP1, C-NAP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11190 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BV73 | |
| CEP250 | |
| CEP250/CNAP1 Fusion Protein Ag5925 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction