missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP350 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555827
This item is not returnable.
View return policy
Description
Immunogen sequence: QDWFDEDFGL SSSHKIQKNK AEETIVPLMA EPKRVTQQPC ETLLAVPHTA EEVEILVHNA AEELWKWKEL GHDLHSISIP TKLLGCASKG LDIESTSKRV YK Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
May be involved in transcriptional regulation.
Specifications
| CEP350 | |
| Polyclonal | |
| Unconjugated | |
| CEP350 | |
| 4933409L06Rik; 6430546F08Rik; Cap350; centrosomal protein 350; centrosomal protein 350kDa; centrosome-associated protein 350; Centrosome-associated protein of 350 kDa; CEP350; GM133; Kiaa0480 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VT06 | |
| CEP350 | |
| Recombinant protein corresponding to Human CEP350. Recombinant protein control fragment (Product #RP-94227). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction