missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP55 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97195R
This item is not returnable.
View return policy
Description
CEP55 Polyclonal antibody specifically detects CEP55 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CEP55 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55165 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| C10orf3, cancer/testis antigen 111, centrosomal protein 55kDa, centrosomal protein of 55 kDa, Cep55, chromosome 10 open reading frame 3, CT111, FLJ10540, Up-regulated in colon cancer 6, URCC6 | |
| Produced in rabbits immunized with E. coli-derived Human CEP55 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction