missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Somatostatin R5/SSTR5 Antibody (394401), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4448H
This item is not returnable.
View return policy
Description
Somatostatin R5/SSTR5 Monoclonal antibody specifically detects Somatostatin R5/SSTR5 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Somatostatin R5/SSTR5 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 6755 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 394401 | |
| Flow Cytometry, CyTOF-ready | |
| somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R | |
| NS0 mouse myeloma cell line transfected with human Somatostatin R5/SSTR5, Met1-Leu364, Accession # AAK61266 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction