missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP68 Antibody (OTI5B4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71728AF700
This item is not returnable.
View return policy
Description
CEP68 Monoclonal specifically detects CEP68 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| CEP68 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CEP68 | |
| Full length recombinant protein of human CEP68 (NP_055962) produced in HEK293T cells. | |
| 0.1 mL | |
| 23177.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI5B4 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| centrosomal protein 68kDa, centrosomal protein of 68 kDa, Cep68, FLJ36750, KIAA0582FLJ25920 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction