missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP72 Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811141S
This item is not returnable.
View return policy
Description
CEP72 Monoclonal antibody specifically detects CEP72 in Human samples. It is validated for Western BlotSpecifications
| CEP72 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15228 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8443 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B8 | |
| Unconjugated | |
| GNPAT | |
| 2610029E11Rik; 4933440J22Rik; centrosomal protein 72; centrosomal protein 72 kDa; centrosomal protein 72kDa; centrosomal protein of 72 kDa; Cep72; KIAA1519 | |
| Full length human recombinant protein of human GNPAT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction