Learn More
Invitrogen™ CEP72 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103730
Product Code. 16610185
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of total CEP72.
Involved in the recruitment of key centrosomal proteins to the centrosome. Provides centrosomal microtubule-nucleation activity on the gamma-tubulin ring complexes (gamma-TuRCs) and has critical roles in forming a focused bipolar spindle, which is needed for proper tension generation between sister chromatids. Required for localization of KIZ, AKAP9 and gamma-tubulin ring complexes (gamma-TuRCs).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEP72
Polyclonal
Unconjugated
CEP72
2610029E11Rik; 4933440J22Rik; centrosomal protein 72; centrosomal protein 72 kDa; centrosomal protein 72kDa; centrosomal protein of 72 kDa; Cep72; KIAA1519
Rabbit
Affinity chromatography
RUO
55722, 74470
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9D3R3, Q9P209
CEP72
A synthesized peptide derived from human CEP72(Accession Q9P209), corresponding to amino acid residues S229-P279.
100 μL
Primary
Human, Mouse, Monkey