missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEP83 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117728
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.
Specifications
| CEP83 | |
| Polyclonal | |
| Unconjugated | |
| CEP83 | |
| 2600001G24Rik; 4921537D05Rik; 5730513H21Rik; CCDC41; centrosomal protein 83; centrosomal protein 83kDa; centrosomal protein of 83 kDa; Cep83; coiled-coil domain containing 41; coiled-coil domain-containing protein 41; NPHP18; NY-REN-58; NY-REN-58 antigen; renal carcinoma antigen NY-REN-58 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51134 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9Y592 | |
| CEP83 | |
| Recombinant Human Centrosomal protein of 83 kDa protein (34-260aa). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction