Learn More
Invitrogen™ CEP83 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121204
Description
Positive test controls include: HT-29, Mouse lung, Mouse spleen, Mouse thymus. The target is usually found in the following locations: Cytoplasm, centriole, centrosome, cytoskeleton, microtubule organizing center. Immunogen sequence: MLIDERLRCE HHKANYQTLK AEHTRLQNEH VKLQNELKHL FNEKQTQQEK LQLLLEELRG ELVEKTKDLE EMKLQILTPQ KLELLRAQIQ QELETPMRER FRNLDEEVEK YRAVYNKLRY EHTFLKSEFE HQKEEYARIL DEGKIKYESE IARLEEDKEE LRNQLLNVDL TKDSKRVEQL AREKVYLCQK LKGLEAEVAE LKAEKENSEA QVENAQRIQV RQLAEMQ.
Component of the distal appendage region of the centriole involved in the initiation of primary cilium assembly. May collaborate with IFT20 in the trafficking of ciliary membrane proteins from the Golgi complex to the cilium during the initiation of primary cilium assembly.
Specifications
| CEP83 | |
| Polyclonal | |
| Unconjugated | |
| CEP83 | |
| 2600001G24Rik; 4921537D05Rik; 5730513H21Rik; CCDC41; centrosomal protein 83; centrosomal protein 83kDa; centrosomal protein of 83 kDa; Cep83; coiled-coil domain containing 41; coiled-coil domain-containing protein 41; NPHP18; NY-REN-58; NY-REN-58 antigen; renal carcinoma antigen NY-REN-58 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51134, 77048 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.73 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D5R3, Q9Y592 | |
| CEP83 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 34-260 of human CEP83 (NP_057206.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.