missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP97 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-06508AF488
This item is not returnable.
View return policy
Description
CEP97 Polyclonal antibody specifically detects CEP97 in Human samples. It is validated for Western Blot
Specifications
| CEP97 | |
| Polyclonal | |
| Western Blot | |
| 2810403B08Rik, centrosomal protein 97kDa, centrosomal protein of 97 kDa, Cep97, FLJ23047, FLJ26462, Leucine-rich repeat and IQ domain-containing protein 2, leucine-rich repeats and IQ motif containing 2, LRRIQ2 | |
| The immunogen recognized by this antibody maps to a region between residue 625 and 675 of human centrosomal protein 97kDa using the numbering given in entry NP_078824.2 (GeneID 79598). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79598 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction