missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEPT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13228388
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13228388 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13228388 Supplier Invitrogen™ Supplier No. PA523876

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, rat and Xenopus based on sequence homology.

Choline phosphotransferase catalyzes the final step in the synthesis of phosphatidylcholine by the transfer of phosphocholine from CDP-choline to diacylglycerol. The synthesis of phosphatidyl ethanolamine by ethanolamine phosphotransferase occurs using an analogous reaction. This gene codes for a choline/ethanolamine phosphotransferase. The protein can synthesize either choline- or ethanolamine- containing phospholipids. Two alternatively spliced transcripts encoding the same isoform have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEPT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CEPT1
Gene Accession No. Q8BGS7, Q9Y6K0
Gene Alias 9930118K05Rik; CEPT1; choline/ethanolamine phosphotransferase 1; choline/ethanolaminephosphotransferase; choline/ethanolaminephosphotransferase 1; hCEPT1; mCEPT1; PRO1101
Gene Symbols CEPT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 29-57 amino acids from the N-terminal region of human CEPT1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10390, 99712
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.