missing translation for 'onlineSavingsMsg'
Learn More

Ceralasertib, MedChemExpress

Product Code. 16490188
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16490188

Brand: Medchem Express HY19323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ceralasertib (AZD6738) is an orally active and bioavailable inhibitor of ATR kinase with an IC50 of 1 nM.

Ceralasertib (AZD6738) is an orally active and bioavailable inhibitor of ATR kinase with an IC50 of 1 nM.

Specifications

Chemical Name or Material Ceralasertib
Physical Form Solid
CAS 1352226-88-0
Color Pink
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H24N6O2S
For Use With (Application) Cancer-Kinase/protease
Synonym AZD6738
Solubility Information DMSO : ≥ 38 mg/mL (92.12 mM)
SMILES O=[S@@](C1(CC1)C2=NC(C3=C4C(NC=C4)=NC=C3)=NC(N5CCOC[C@H]5C)=C2)(C)=N
Molecular Weight (g/mol) 412.51
Formula Weight 412.51
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.