missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ceramide Kinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30872-25ug
This item is not returnable.
View return policy
Description
Ceramide Kinase Polyclonal antibody specifically detects Ceramide Kinase in Human samples. It is validated for ELISA,Western Blot
Specifications
| Ceramide Kinase | |
| Polyclonal | |
| Western Blot : 1-10 μg/mL, ELISA | |
| ceramide kinase, dA59H18.2, DKFZp434E0211, EC 2.7.1.138, FLJ21430, FLJ23239, hCERKLipid kinase 4, KIAA1646dA59H18.3, lipid kinase LK4, LK4Acylsphingosine kinase, MGC131878 | |
| Rabbits were immunized with recombinant full-length human Ceramide Kinase protein | |
| 25 μg | |
| Apoptosis, Cell Biology, Lipid and Metabolism | |
| 64781 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction