missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerberus 1 Antibody (034), DyLight 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89662UV
This item is not returnable.
View return policy
Description
Cerberus 1 Monoclonal antibody specifically detects Cerberus 1 in Human samples. It is validated for ELISA
Specifications
| Cerberus 1 | |
| Monoclonal | |
| DyLight 350 | |
| cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Cerberus 1 (AAH69371.1; Met1-Ala267). | |
| 0.1 mL | |
| Stem Cell Markers, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 9350 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 034 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction