missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerebellin-1/Precerebellin Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77238H
This item is not returnable.
View return policy
Description
Cerebellin-1/Precerebellin Polyclonal antibody specifically detects Cerebellin-1/Precerebellin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| Cerebellin-1/Precerebellin | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| cerebellin 1 precursor, cerebellin-1, CLN1, precerebellin | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy- terminus human precerebellin. The immunogen is located within the last 50 amino acids of Precerebellin. Amino Acid Squence: QMEKGDRAYLKLERGN | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 869 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction