Learn More
Tocris Bioscience™ Cerestat
Potent and noncompetitive NMDA antagonist
Brand: Tocris Bioscience™ 4492/50
Description
Cerestat is a potent and noncompetitive NMDA receptor antagonist (IC50 = 36 nM). Exhibits >70-fold selectivity for NMDA receptor over σ receptor. Neuroprotective in a rat model of focal ischemia.Specifications
| Cerestat | |
| N-(3-Ethylphenyl)-N-methyl-N'-1-naphthalenylguanidine hydrochloride | |
| Soluble to 50mM in water and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 137160-11-3 | |
| 50 mg | |
| Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research Use Only. Not For Human or Veterinary Use