missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CERK Polyclonal Antibody
GREENER_CHOICE

Product Code. 17149503
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17149503 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17149503 Supplier Invitrogen™ Supplier No. PA5111586

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VLHGLIGRTQ RSAGVDQNHP RAVLVPSSLR IGIIPAGSTD CVCYSTVGTS DAETSALHIV VGDSLAMDVS SVHHNSTLL Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 89%.

CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CERK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CERK
Gene Accession No. Q8TCT0
Gene Alias acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646
Gene Symbols CERK
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human CERK. Recombinant protein control fragment (Product #RP-105333).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.