missing translation for 'onlineSavingsMsg'
Learn More

CERT Polyclonal Antibody
GREENER_CHOICE

Product Code. 17207803 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17207803 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17207803 Supplier Thermo Scientific Supplier No. PA5115035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CERT Polyclonal Antibody

CERT Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CERT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene CERT1
Gene Accession No. Q9EQG9, Q9Y5P4
Gene Alias 2810404O15Rik; 9230101K08Rik; alpha 3 type IV collagen binding protein; AU016711; Ceramide transfer protein; ceramide transport protein; ceramide transporter; Cert; CERTL; Col4a3bp; collagen type IV alpha 3 binding protein; collagen type IV alpha-3-binding protein; collagen, type IV, alpha 3 (Goodpasture antigen) binding protein; goodpasture antigen-binding protein; GPBP; hCERT; lipid-transfer protein CERTL; MRD34; procollagen, type IV, alpha 3 (Goodpasture antigen) binding protein; STARD11; StAR-related lipid transfer (START) domain containing 11; stAR-related lipid transfer protein 11; START domain-containing protein 11
Gene Symbols CERT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human COL4A3BP(Accession Q9Y5P4), corresponding to amino acid residues K284-D334.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10087, 365652, 68018
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.