missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ceruloplasmin Antibody (8F3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42940
This item is not returnable.
View return policy
Description
Ceruloplasmin Monoclonal antibody specifically detects Ceruloplasmin in Human,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Ceruloplasmin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 8F3 | |
| Western Blot 1:500 - 1:5000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| ceruloplasmin, ceruloplasmin (ferroxidase), CP-2, EC 1.16.3.1, Ferroxidase | |
| ceruloplasmin(ferroxidase) - (Uniprot# P00450) | |
| 100 μg | |
| Cardiovascular Biology, Neurodegeneration, Neuroscience | |
| 1356 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction