missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CESK1 Antibody (OTI1C1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46114
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
CESK1 Monoclonal specifically detects CESK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| CESK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| CESK1MGC118839, chaperonin containing TCP1, subunit 8 (theta)-like 2, MGC118840, putative T-complex protein 1 subunit theta-like 2, T-complex protein 1 | |
| Mouse | |
| 59.2 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1C1 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| CCT8L2 | |
| Full length human recombinant protein of human CCT8L2(NP_055221) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| 150160 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction