missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CETN1 Monoclonal Antibody (OTI1B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804225
This item is not returnable.
View return policy
Description
The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This encoded protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.Specifications
| CETN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12798 | |
| CETN1 | |
| Full length human recombit protein of human CETN1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B12 | |
| Unconjugated | |
| CETN1 | |
| CEN1, CETN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1068 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction