Learn More
CETN1 Mouse anti-Human, Clone: OTI7G7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand:  Origene Technologies CF804388
Encompass_Preferred
Product Code. 17687961
  • POA / 100µg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene plays important roles in the determination of centrosome position and segregation, and in the process of microtubule severing. This encoded protein is localized to the centrosome of interphase cells, and redistributes to the region of the spindle poles during mitosis, reflecting the dynamic behavior of the centrosome during the cell cycle.
Specifications
Specifications
CETN1
Monoclonal
Unconjugated
CETN1
CEN1, CETN
Mouse
Affinity Chromatography
RUO
1068
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Western Blot
OTI7G7
PBS with 8% trehalose and no preservative; pH 7.3
Q12798
CETN1
Full length human recombit protein of human CETN1 produced in E.coli.