missing translation for 'onlineSavingsMsg'
Learn More

CETN3 Monoclonal Antibody (OTI4A10), TrueMAB™, OriGene

Product Code. 17505913
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17505913 30 μL 30µL
17585903 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17505913 Supplier Origene Technologies Supplier No. TA805891S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CETN3 Monoclonal antibody specifically detects CETN3 in Human samples. It is validated for Western Blot

Specifications

Antigen CETN3
Applications Western Blot
Classification Monoclonal
Clone OTI4A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cetn3
Gene Accession No. O15182
Gene Alias CDC31; CDC31 yeast homolog; CEN3; centrin 3; centrin, EF-hand protein, 3; centrin, EF-hand protein, 3 (CDC31 homolog, yeast); centrin-3; Cetn3; EF-hand superfamily member; MmCEN3
Host Species Mouse
Immunogen Full length human recombinant protein of human CETN3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1070
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.