missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cetuximab Antibody (2259C) [Biotin], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB9626B
This item is not returnable.
View return policy
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab in Broad Species Reactivity samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA
Specifications
| Cetuximab | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Full length Cetuximab, | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Simple Western, Flow Cytometry, ELISA | |
| 2259C | |
| Western Blot, Simple Western, Flow Cytometry, ELISA | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction