Learn More
Cetuximab Mouse, Biotin, UltraLys, Monoclonal Antibody, AcroBiosystems
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab, recommended for PK/PD.
Specifications
Specifications
| Antigen | Cetuximab |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Host Species | Hybridoma |
| Quantity | 25 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | -20°C |