Learn More
Cetuximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Product Code. 16417419
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417419 100 μg 100µg
16427419 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16417419 Supplier ACROBiosystems Supplier No. CEBY31100ug
Mouse Monoclonal Antibody
Cetuximab Monoclonal antibody specifically detects Cetuximab (Non-Neutralizing)
Specifications
Antigen Cetuximab
Classification Monoclonal
Conjugate Unconjugated
Host Species Hybridoma
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C