Learn More
Cetuximab Mouse, Unconjugated, Monoclonal Antibody, AcroBiosystems
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab (Non-Neutralizing)
Specifications
Specifications
| Antigen | Cetuximab |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Hybridoma |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Content And Storage | -20°C |