missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP299 Antibody (OTI1A6), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72466AF532
This item is not returnable.
View return policy
Description
CFAP299 Monoclonal specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CFAP299 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CFAP299 | |
| Full length recombinant protein of human C4orf22 (NP_689983) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry | |
| chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
| Mouse | |
| 0.1 mL | |
| 255119 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido