missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP299 Antibody (OTI1A6), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72466JF646
This item is not returnable.
View return policy
Description
CFAP299 Monoclonal specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CFAP299 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CFAP299 | |
| Full length recombinant protein of human C4orf22 (NP_689983) produced in HEK293T cells. | |
| 0.1 mL | |
| 255119.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry | |
| chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction