missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP299 Antibody (OTI1A6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72466PCP
This item is not returnable.
View return policy
Description
CFAP299 Monoclonal antibody specifically detects CFAP299 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CFAP299 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 255119 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry | |
| chromosome 4 open reading frame 22, hypothetical protein LOC255119, MGC35043 | |
| Full length human recombinnat protein of human C4orf22(NP_689983) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction