missing translation for 'onlineSavingsMsg'
Learn More

Perforin Antibody (CE2.10), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97440UV

Product Code. 18016637

  • ONLINE EXCLUSIVE PRICE
    € 693.00
    € 655.20 / 0.10mL
    Save € 37.80
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Perforin Monoclonal specifically detects Perforin in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Perforin
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
PRF1
Recombinant mouse perforin (aa 98-534).
0.1 mL
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity
Recognizes an epitope in the region aa 402-534 of human and mouse perforin. Does NOT detect Human in Western Blot.
Store at 4°C in the dark.
Western Blot
CE2.10
Western Blot
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.