Learn More
Invitrogen™ CFAP97 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5114106
Product Code. 17161964
  • € 506.00 / 100µL
Description
Immunogen sequence: NMGYLNSSPL SRRARSTLGQ YSPLRASRTS SATSGLSCRS ERSAVDPSSG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 80%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 87%; Rabbit: 93%; Rat: 93%.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CFAP97
Polyclonal
Unconjugated
CFAP97
CFAP97; cilia and flagella associated protein 97; cilia- and flagella-associated protein 97; hmw; KIAA1430
Rabbit
Affinity Chromatography
RUO
57587
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9P2B7
CFAP97
Synthetic peptide directed towards the middle region of human KIAA1430.