missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CFAP97D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144729
This item is not returnable.
View return policy
Description
Immunogen sequence: MNNSLDYLAY PVIVSNHRQS TTFRKKLDFG HYVSHKNRIQ IAKPTVDTKP PVAHTNHILK LSKLQGEQKK INKIEYENKQ L Antigen sequence identity: Mouse 85%; Rat 85%.
CFAP97D1 is a protein that is encoded by the CFAP97D1 gene in humans. It is a component of the cilia and flagella, which are hair-like structures that protrude from the surface of cells and are involved in various cellular processes, including movement and signaling.
Specifications
| CFAP97D1 | |
| Polyclonal | |
| Unconjugated | |
| C17orf105 | |
| C17orf105; CFAP97 domain-containing protein 1; CFAP97D1; chromosome 17 open reading frame 105; uncharacterized protein C17orf105; Uncharacterized protein CFAP97D1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 284067 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| B2RV13 | |
| C17orf105 | |
| Recombinant protein fragment of Human C17orf105 (Product #RP-103144) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction