missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFH Monoclonal Antibody (OTI5H5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804532S
This item is not returnable.
View return policy
Description
CFH Monoclonal antibody specifically detects CFH in Human samples. It is validated for Western BlotSpecifications
| CFH | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q2MKA7 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 284654 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI5H5 | |
| Unconjugated | |
| RSPO1 | |
| adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1 | |
| Human recombinant protein fragment corresponding to amino acids 21-263 of human RSPO1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction